missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SHBG Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00€ - 377.00€
Specifications
| Antigen | SHBG |
|---|---|
| Clone | 336729 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18762758
![]() |
R&D Systems
MAB26562-SP |
25 μg |
188.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18790339
![]() |
R&D Systems
MAB26562-100 |
100 μg |
377.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHBG Monoclonal specifically detects SHBG in Human samples. It is validated for ELISA.Specifications
| SHBG | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ABP, ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin | |
| Mouse myeloma cell line NS0-derived recombinant human SHBG Leu30-His402 Accession # P04278 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human SHBG in direct ELISAs. |
| 336729 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P04278 | |
| 6462 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title