missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human SLC31A1 Alexa Fluor™ 750-conjugated Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ FAB11484S-100UG
This item is not returnable.
View return policy
Description
SLC31A1/CTR1 Monoclonal antibody specifically detects SLC31A1/CTR1 in Human samples. It is validated for Flow Cytometry
Specifications
| SLC31A1/CTR1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| O15431 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 1317 | |
| Protect from light.Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG |
| Flow Cytometry | |
| 2938A | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Copper transporter 1, COPT1, COPT1high affinity copper uptake protein 1, CTR1, CTR1MGC75487, HCTR1, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1 | |
| Synthetic peptide, Accession # O15431 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction