missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human SLC31A1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ MAB11484-SP
This item is not returnable.
View return policy
Description
SLC31A1/CTR1 Monoclonal antibody specifically detects SLC31A1/CTR1 in Human samples. It is validated for Flow Cytometry
Specifications
| SLC31A1/CTR1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Copper transporter 1, COPT1, COPT1high affinity copper uptake protein 1, CTR1, CTR1MGC75487, HCTR1, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1 | |
| Synthetic peptide, Accession # O15431 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry | |
| 2938A | |
| Flow Cytometry 0.25 μg/106 cells | |
| O15431 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 1317 | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto