Learn More
R&D Systems™ Human Sphingosine Kinase 1 (NP_068807.2) VersaClone cDNA
Specifications
Specifications
| Gene | Sphingosine Kinase 1/SPHK1 |
| Accession Number | NP_068807.2 |
| Gene ID (Entrez) | 8877 |
| Gene Alias | EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |