missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPTLC2 Synthetic Peptide

Product Code. 15235259
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15235259 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15235259 Supplier Invitrogen™ Supplier No. PEP1256
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 15 amino acids near the carboxy terminus of human SPT2. PEP-1256 can be used as a blocking peptide with polyclonal antibody PA5-21142.

Serine palmitoyltransferase (SPT), which consists of two different subunits, is the key enzyme in sphingolipid biosynthesis. It converts L-serine and palmitoyl-CoA to 3-oxosphinganine with pyridoxal 5'-phosphate as a cofactor. SPT2 is the long chain base subunit 2 of mammalian serine palmitoyltransferase. SPT2 is catalytically active but needs its related protein SPT1 for its stabilization and anchoring the holoenzyme to the cytosolic face of the endoplasmic reticulum. As in the case with SPT1, mutations in the SPT2 gene can cause hereditary sensory and autonomic neuropathy type I (HSAN-I), resulting from a loss of SPT activity as well as the accumulation of the atypical and neurotoxic sphingoid metabolite 1-deoxy-sphinganine.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9517
Species Human
Purification Method purified
Accession Number O15270
Common Name SPTLC2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias AI173915; hLCB2a; HSN1C; KIAA0526; LCB 2; LCB2; LCB2a; long chain base biosynthesis protein 2; Long chain base biosynthesis protein 2a; mKIAA0526; NSAN1C; Pomt2; protein-O-mannosyltransferase 2; serine palmitoyltransferase 2; serine palmitoyltransferase long chain base subunit 2; serine palmitoyltransferase, long chain base subunit 2; serine palmitoyltransferase, subunit II; serine-palmitoyl-CoA transferase 2; SPT 2; SPT2; Sptlc2
Gene Symbol SPTLC2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.