Learn More
Human Sulfamidase/SGSH Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8380-SP
This item is not returnable.
View return policy
Description
Sulfamidase/SGSH Monoclonal specifically detects Sulfamidase/SGSH in Human samples. It is validated for Immunocytochemistry.
Specifications
| Sulfamidase/SGSH | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| heparan sulfate sulfatase, HSS, HSSN-sulphoglucosamine sulphohydrolase, MPS3A, MPS3Asulphamidase, N-sulfoglucosamine sulfohydrolase, SFMD, SFMDEC 3.10.1.1, SGSH, Sulfoglucosamine sulfamidase, Sulphamidase | |
| Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Sulfamidase/SGSH protein, Arg23-Leu502, Accession # P51688 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunocytochemistry | |
| 1018387 | |
| Immunocytochemistry 8-25 ug/mL | |
| P51688 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6448 | |
| Human | |
| Purified |