missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sulfatase Modifying Factor 2/SUMF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3594-SP
This item is not returnable.
View return policy
Description
Sulfatase Modifying Factor 2/SUMF2 Monoclonal specifically detects Sulfatase Modifying Factor 2/SUMF2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Sulfatase Modifying Factor 2/SUMF2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q8NBJ7 | |
| SUMF2 | |
| Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2 Gln26-Leu301 Accession # Q8NBJ7 | |
| 25 μg | |
| Primary | |
| Detects human Sulfatase Modifying Factor 2/SUMF2 in direct ELISAs and Western blots. No cross-reactivity with recombinant human SUMF1 or recombinant mouse SUMF2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 330312 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-alpha-formyglycine-generating enzyme 2, C-alpha-formylglycine-generating enzyme 2, DKFZp566I1024, DKFZp686I1024, DKFZp686L17160, DKFZp781L1035, FGE2, MGC99485, paralog of the formylglycine-generating enzyme, pFGE, sulfatase modifying factor 2, sulfatase-modifying factor 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 25870 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction