Learn More
Human Syndecan-2/CD362 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems FAB2965S-100UG
Product Code. 18721609
  • 532.00€ / 100µg
Description
Syndecan-2/CD362 Monoclonal specifically detects Syndecan-2/CD362 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Syndecan-2/CD362
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
AAH49836.1
SDC2
Mouse myeloma cell line NS0-derived recombinant human Syndecan-2/CD362 Glu19-Gly144 Accession # AAH49836.1
RUO
6383
Human
IgG2b
Flow Cytometry
305515
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD362, CD362 antigen, Fibroglycan, heparan sulfate proteoglycan 1, cell surface-associated, Heparan sulfate proteoglycan core protein, HSPG, HSPG1, HSPG1syndecan proteoglycan 2, SDC2, SYND2, SYND2cell surface-associated heparan sulfate proteoglycan 1, syndecan 2, syndecan-2, Syndecan2
Rat
100 μg
Primary
Detects human Syndecan-2/CD362 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Syndecan-1, recombinant human (rh) Syndecan-3, or rhSyndecan-4 is observed.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
For Research Use Only (RUO)