missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TDO2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC9768N-100UG
This item is not returnable.
View return policy
Description
TDO2 Monoclonal specifically detects TDO2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TDO2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| chky, TDO, TDOTO, TO, TPH2EC 1.13.11.11, TRPO, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan Oxygenase, Tryptophan Pyrrolase, Tryptophanase | |
| E. coli-derived human TDO2, Leu18-Phe388, Accession # P48775 | |
| 100 μg | |
| 6999 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 998604 | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| P48775 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human TDO2 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction