missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-9 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB4091-SP
This item is not returnable.
View return policy
Description
IL-9 Monoclonal specifically detects IL-9 in Mouse samples. It is validated for Neutralization.
Specifications
| IL-9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytokine P40, HP40, IL9, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3578 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Neutralization | |
| 222622 | |
| Neutralization 0.5-2.0 ng/mL | |
| P15247.1 | |
| IL9 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-9 Gln19-Pro144 Accession # P15247.1 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-9 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human IL-9. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction