missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LRRC32/GARP Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB62291N-100UG
This item is not returnable.
View return policy
Description
LRRC32/GARP Monoclonal specifically detects LRRC32/GARP in Mouse samples. It is validated for Flow Cytometry.
Specifications
| LRRC32/GARP | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| D11S833E, D11S833Eleucine-rich repeat-containing protein 32, GARP, GARPGarpin, Garpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse LRRC32/GARP in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) LRRC32 is observed and no cross-reactivity with rhLRRC3, rhLRRC4, or rhNGL-3/LRRC4B is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 725226 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| NP_001106850 | |
| LRRC32 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse LRRC32/GARP, Ile18-Asn628, Accession # NP_001106850 | |
| 100 μg | |
| 2615 | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction