missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti- Brentuximab (Anti-Idiotype) Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB9584S-100UG
This item is not returnable.
View return policy
Description
Brentuximab Monoclonal specifically detects Brentuximab in Multi-Species samples. It is validated for Flow Cytometry.
Specifications
| Brentuximab | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Full length Brentuximab | |
| 100 μg | |
| Primary | |
| Multi-species | |
| Purified |
| Flow Cytometry | |
| 2262C | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects Brentuximab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Trastuzumab, Tositumomab, Adalimumab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction