missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TLR4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
247.00€ - 551.00€
Specifications
| Antigen | TLR4 |
|---|---|
| Clone | 1039412 |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18750347
![]() |
R&D Systems
MAB11187-SP |
25 μg |
247.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18720907
![]() |
R&D Systems
MAB11187-100 |
100 μg |
551.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR4 Monoclonal antibody specifically detects TLR4 in Human samples. It is validated for Western BlotSpecifications
| TLR4 | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ARMD10, CD_antigen: CD284, CD284, CD284 antigen, EC 3.2.2.6, EC:3.2.2.6, homolog of Drosophila toll, hToll, TLR4, TLR-4, TOLL, toll like receptor 4 protein, toll-like receptor 4 | |
| Mouse myeloma cell line NS0-derived human TLR4, Glu24-Lys631, Accession # O00206 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1039412 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O00206 | |
| 7099 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title