missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TMPRSS11B (NP_872308.2) VersaClone cDNA

Product Code. 18762207
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18762207 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18762207 Supplier R&D Systems™ Supplier No. RDC3506

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene TMPRSS11B
Accession Number NP_872308.2
Gene ID (Entrez) 132724
Gene Alias Airway trypsin-like protease 5, DKFZp686L1818, EC 3.4.21.-, HATL5, transmembrane protease serine 11 B, transmembrane protease, serine 11 B
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.