missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TR2/NR2C1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems PP-H0037-00
This item is not returnable.
View return policy
Description
TR2/NR2C1 Monoclonal specifically detects TR2/NR2C1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Direct ELISA.
Specifications
| TR2/NR2C1 | |
| Monoclonal | |
| Unconjugated | |
| A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
| NR2C1 | |
| Recombinant human TR2/NR2C1 aa 4-110 | |
| 100 μg | |
| Primary | |
| This antibody specifically recognizes human TR2. Not yet tested in other species. | |
| This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunoprecipitation, ELISA | |
| H0037 | |
| Western Blot, Immunoprecipitation, Direct ELISA | |
| NR2C1, nuclear receptor subfamily 2 group C member 1, nuclear receptor subfamily 2, group C isoform, nuclear receptor subfamily 2, group C, member 1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2, TR2 nuclear hormone receptor, TR2-11 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7181 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction