missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Transcobalamin II Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7895-SP
This item is not returnable.
View return policy
Description
Transcobalamin II Polyclonal specifically detects Transcobalamin II in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Transcobalamin II | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| D22S676, D22S750, macrocytic anemia, TC, TC-2, TC2II, TCII, TCN2, transcobalamin II; macrocytic anemia, transcobalamin IITC II, transcobalamin-2, vitamin B12-binding protein 2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6948 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P20062 | |
| TCN2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Transcobalamin II Glu19-Trp427 Accession # P20062 | |
| 25 μg | |
| Primary | |
| Detects human Transcobalamin II in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction