missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Transglutaminase 7/TGM7 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5426-SP
This item is not returnable.
View return policy
Description
Transglutaminase 7/TGM7 Polyclonal specifically detects Transglutaminase 7/TGM7 in Human samples. It is validated for Western Blot.
Specifications
| Transglutaminase 7/TGM7 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TGM7 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Transglutaminase 7/TGM7 | |
| 25 μg | |
| Primary | |
| Detects human Transglutaminase 7/TGM7 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Transglutaminase 3 is observed, and less than 2% cross-reactivity with rhTransglutaminase 2, rhTransglutaminase 4, and recombinant mouse Transglutaminase 2 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| protein-glutamine gamma-glutamyltransferase Z, TG(Z), TGase Z, TGase-7, TGM7, TGMZ, TGMZEC 2.3.2.13, TGZ, transglutaminase 7, Transglutaminase Z, transglutaminase-7 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 116179 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction