Learn More
R&D Systems™ Human TRIM9 (NP_055978.4) VersaClone cDNA
Specifications
Specifications
| Accession Number | NP_055978.4 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |