Learn More
Human ULBP-3 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 6 publications
Brand:  R&D Systems AF1517-SP
 View more versions of this product
Product Code. 18707367
  • 209.00€ / 25µg
Description
ULBP-3 Polyclonal specifically detects ULBP-3 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ULBP-3
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
NKG2D ligand 3, RAET1N, UL16 binding protein 3, ULBP3
Goat
Affinity Purified
RUO
79465
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Blockade of Receptor-ligand Interaction
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
NP_078794
ULBP3
Mouse myeloma cell line NS0-derived recombinant human ULBP-3 Gly27-Pro216 Accession # NP_078794
25 μg
Primary
Detects human ULBP-3 in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human (rh) ULBP-2 is observed and less than 1% cross-reactivity with rhULBP-1 is observed.