missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human UNC5H3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1005-SP
This item is not returnable.
View return policy
Description
UNC5H3/UNC5C Monoclonal specifically detects UNC5H3/UNC5C in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| UNC5H3/UNC5C | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| O95185 | |
| UNC5C | |
| Mouse myeloma cell line NS0-derived recombinant human UNC5H3 Ala40-Asp376 Accession # O95185 | |
| 25 μg | |
| Primary | |
| Detects human UNC5H3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human UNC5H4 or recombinant rat UNC5H2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 200414 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| netrin receptor UNC5C, Protein unc-5 homolog 3, Protein unc-5 homolog C, unc-5 homolog 3, unc-5 homolog C (C. elegans), UNC5C, UNC5H3unc5 (C.elegans homolog) c | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8633 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction