missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human USP19 (NP_006668) VersaClone cDNA
Description
The Human USP19 (NP_006668) VersaClone cDNA clone contains the complete ORF for USP19, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The USP19 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Spécification
Spécification
| Product Type | cDNA |
| Gene | USP19 |
| Accession Number | NP_006668 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Gene ID (Entrez) | 10869 |
| Gene Alias | Deubiquitinating enzyme 19, EC 3.1.2.15, EC 3.4.19.12, KIAA0891, Ubiquitin Carboxyl-Terminal Hydrolase 19, ubiquitin specific peptidase 19, ubiquitin thioesterase 19, Ubiquitin thiolesterase 19, Ubiquitin-specific-processing protease 19, Zinc finger MYND domain-containing protein 9, ZMYND9, ZMYND9ubiquitin specific protease 19 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
For Research Use Only
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?