missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Uteroglobin/SCGB1A1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4218-SP
This item is not returnable.
View return policy
Description
Uteroglobin/SCGB1A1 Polyclonal specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Immunohistochemistry, Neutralization.
Specifications
| Uteroglobin/SCGB1A1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| blastokinin, CC10, CC10Urine protein 1, CC16, CCPBP, CCSP, CCSPCCPBP, Clara, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, PCB-BP, SCGB1A1, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGB, UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7356 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry, Neutralization | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL, Neutralization 0.3-1.2 ug/mL | |
| NP_003348 | |
| SCGB1A1 | |
| E. coli-derived recombinant human Uteroglobin/SCGB1A1 Glu22-Asn91 Accession # NP_003348 | |
| 25 μg | |
| Primary | |
| Detects human Uteroglobin/SCGB1A1 in direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction