missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGFR3/Flt-4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 12 publications
Brand: R&D Systems AF349
This item is not returnable.
View return policy
Description
VEGFR3/Flt-4 Polyclonal specifically detects VEGFR3/Flt-4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| VEGFR3/Flt-4 | |
| Polyclonal | |
| Unconjugated | |
| P35916 | |
| FLT4 | |
| Mouse myeloma cell line NS0-derived recombinant human VEGFR3/Flt-4, Tyr25-Ile776, Accession # P35916 | |
| 100 μg | |
| Primary | |
| Detects human VEGFR3/Flt-4 in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse VEGFR3 is observed and less than 2% cross-reactivity with recombinant human VEGFR1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| EC 2.7.10, EC 2.7.10.1, Flt-4, FLT4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGF R3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2324 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction