missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VPS34/PIK3C3 (NP_002638) VersaClone cDNA

Product Code. 18707489
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18707489 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18707489 Supplier R&D Systems™ Supplier No. RDC2564
  • ONLINE EXCLUSIVE PRICE
    761.00€
    719.25€ / 10µg
    Save 41.75€
    5% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • PIK3C3 phosphatidylinositol 3- kinase catalytic subunit type 3 [ Homo sapiens (human) ]
  • Also known as: VPS34; Vps34; hVps34
  • The VPS34 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • PIK3C3 is a catalytic subunit of the PI3K complex that mediates formation of phosphatidylinositol 3- phosphate.
  • Different complex forms are believed to play a role in multiple membrane trafficking pathways.
  • PI3KC3- C1 is involved in initiation of autophagosomes and PI3KC3- C2 in maturation of autophagosomes and endocytosis.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene VPS34
Accession Number NP_002638
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 2677 bp
Gene ID (Entrez) 5289
Gene Alias EC 2.7.1, EC 2.7.1.137, hVps34, MGC61518, phosphatidylinositol 3-kinase catalytic subunit type 3, Phosphatidylinositol 3-kinase p100 subunit, Phosphoinositide-3-kinase class 3, phosphoinositide-3-kinase, class 3, PI 3-Kinase C3, PI3K type 3, PI3-kinase type 3, PIK3C3, PtdIns-3-kinase type 3, Vps34
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.