missing translation for 'onlineSavingsMsg'
Learn More

Testo™ Huminator II Transport Trolley

Product Code. 15836341
Change view
Click to view available options
Color:
Dark Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15836341 Dark Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15836341 Supplier Testo™ Supplier No. 0519 0899
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Huminator II accessory

The optimal protection for the Huminator II is given by the optionally available transport trolley, promising excellent handling during the on-site use

Specifications

Color Dark Gray
For Use With (Equipment) Huminator II precision humidity generator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.