missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HUNK, GST Tag Recombinant Protein
This HUNK Recombinant Human Protein (full length) was expressed in insect cells.
Brand: Invitrogen™ A33377
Additional Details : Weight : 0.00900kg
Description
HUNK serine/threonine kinase or hormonally up-regulated Neu-associated kinase is a member of the mitogen-activated protein kinase family which is essential for mammary tumor metastasis. HUNK deletion in mice did not alter viability, fertility, longevity, or propensity to develop tumors and it is also dispensable for murine development.- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Specifications
NP_055401 | |
105.9. kDa | |
CAMKL Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
1 mg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Baculovirus | |
Full Length, Recombinant, Active |