Learn More
Cytiva HyClone™ Cell Boost™ 6 Supplement
Product Code. 16605805
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16605805 1000 mL 1 L
16554581 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16605805 Supplier Cytiva Supplier No. SH31118.02
  • ONLINE EXCLUSIVE PRICE
    258.76€
    224.00€ / 1 L
    Save 34.76€
    13% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Fed-batch supplement with lipids, amino acids, vitamins, and growth factors for hybridoma, HEK 293, NS0, CHO, and T cells
  • Chemically Defined (CD), Animal-Derived Component-Free (ADCF), Protein-Free, and Hydrolysate-Free
  • Supports fed-batch cultures with lipids, amino acids, vitamins and growth factors
  • Developed for recombinant protein production with various cell lines including Chinese Hamster Ovary (CHO) cell line, Human Embryonic Kidney (HEK) 293, NS0, hybridoma, and T cells
  • Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
  • Manufactured to meet cGMP manufacturing standards and QC specifications
  • Available as a ready-to-use liquid feed solution
TRUSTED_SUSTAINABILITY
Specifications
Cell Line CHO, HEK293, Hybridoma, NS0, T cells
Form Liquid
Product Type Cell Boost Supplement
Format Bottle
For Use With (Application) Improving Productivity and Cell Yield in Fed-Batch Cultures
Product Line HyClone
Quantity 1000 mL