Learn More
Cytiva HyClone™ Donor Equine Serum (U.S. origin)
Product Code. 17246896
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated
Irradiated (25 to 40 kGy)
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing Unit Size
17246896 500 mL Irradiated (25 to 40 kGy) 1 L
12339822 500 mL Irradiated 500mL
12694207 100 mL Heat Inactivated 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17246896 Supplier Cytiva Supplier No. SH30074.03IR2540
  • ONLINE EXCLUSIVE PRICE
    109.00€
    78.12€ / 1 L
    Save 30.88€
    28% Off
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care. Complete traceability back to the original source.
  • Donor herds are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration
  • The horses are bled in an enclosed facility that is specially equipped for aseptic collection
  • Coggins tests for equine infectious anemia are routinely performed on each horse
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
  • TRUSTED_SUSTAINABILITY
    Specifications
    Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
    Content And Storage Store at or Below -10°C
    Species Equine
    Age Adult
    Country of Origin United States
    Form Frozen
    Format Bottle
    Product Type Equine Serum
    Quantity 500 mL
    Serum Treatment Gamma-irradiated
    Sterility Sterile-filtered
    Sterilization Method Sequential-filtered, 100 nm
    Processing Irradiated (25 to 40 kGy)
    Tested For Equine Herpesvirus, Equine Viral Arteritis, Bovine Virus Diarrhea, Rabies, Reovirus, Cytopathogenic Agnets (IBR), Hemabsorbing Agent (PI3), Equine Infectious Anemia
    Heat Inactivated No
    Product Line HyClone
    Shipping Condition Frozen
    Gamma-irradiated Yes