missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ ES Screened Characterized Fetal Bovine Serum, US Origin
Description
ES Screened FBS supports the growth of undifferentiated murine embryonic stem cells. The screening includes plating efficiency, colony morphology, and toxicity tests.
Specifications
Specifications
| Country of Origin | United States |
| Form | Liquid |
| Specialty Type | ES cell screened |
| Quantity | 500 mL |
| Sterilization Method | 40 nm filtration |
| Processing | Heat Inactivated |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Heat Inactivated | Yes |
| Hemoglobin Concentration | ≤ 10 mg/dL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?