Learn More
Cytiva HyClone HyPure Water for Injection (WFI) Quality Water
Product Code. 17642085
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
2000 mL
6 x 1000 mL
Unit Size:
1 L
100mL
2 L
500mL
6 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17642085 1000 mL 1 L
17622085 100 mL 100mL
17632085 500 mL 500mL
17817697 2000 mL 2 L
17652085 6 x 1000 mL 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17642085 Supplier Cytiva Supplier No. SH31191.03
  • ONLINE EXCLUSIVE PRICE
    35.51€
    30.00€ / 1 L
    Save 5.51€
    15% Off
 Restricted Item
In Stock
Add to basket
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Support cell culture growth and biomanufacturing processes with Water for Injection (WFI) quality water.
  • Support cell culture growth and biomanufacturing processes with Water for Injection (WFI) quality water.
    • Meets stringent USP specifications.
    • Manufactured in ISO 9001 certified facility.
    • Lot-specific certificate of analysis (CoA) for each lot of water produced.
    • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • HyClone HyPure WFI Quality Water undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP). Water for injection (WFI) is produced in ISO 9001 certified facilities to ensure a high quality product.
  • HyPure WFI Quality Water meets the following criteria:
    • Must be produced from a validated water purification system that meets or exceeds current USP requirements for HyPure WFI Quality Water.
    • Must have a quality assurance monitoring system that ensures continuous compliance and incorporates system test and action limits.
    • Must meet all applicable chemical, purity, and other acceptance criteria as outlined in the current USP for both system and packaged water.
    • Must include a lot-specific CoA for each lot of water produced, listing both acceptance criteria and actual test results.
    • Actual test results on a CoA are not taken from system samples, but must be from actual packaged samples of the given lot.
TRUSTED_SUSTAINABILITY
Spezifikation
Sterility Sterile-filtered
Quantity 1000 mL
Physical Form Liquid
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.