Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum, US Origin
Product Code. 12654237
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12654237 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12654237 Supplier Cytiva Supplier No. SH30118.04IR
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Augments cell culture growth, sourced from animals that are typically less than 10 days old
TRUSTED_SUSTAINABILITY
Specifications
Age <10 days
Country of Origin United States
Product Type Calf Bovine Serum
Quantity 1000 mL
Processing Irradiated
For Use With (Application) Augmenting Cell Culture Growth
Product Line HyClone