Learn More
HyClone™ Water, Molecular Biology Grade, Cytiva
Product Code. 10275262
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1 L
Unit Size:
1000mL
100mL
500mL
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10275262 100 mL 100mL
10787944 500 mL 500mL
10307052 1000 mL 1000mL
10247783 6 x 500 mL Pack of 6
10452915 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10275262 Supplier HyClone products Cytiva Supplier No. SH30538.01
  • ONLINE EXCLUSIVE PRICE
    6.50€
    16.00€ / 100mL
    Save POA
    -146.15% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.
  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL