missing translation for 'onlineSavingsMsg'
Learn More

Hydrastinine, TRC

Product Code. 30390792
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390792 25 mg -
30405778 10 mg -
30407186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390792 Supplier Toronto Research Chemicals Supplier No. TRCH67570525MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6592-85-4
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
Synonym 5,6,7,8-Tetrahydro-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-ol
IUPAC Name 6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-ol
SMILES CN1CCc2cc3OCOc3cc2C1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H13NO3/c1-12-3-2-7-4-9-10(15-6-14-9)5-8(7)11(12)13/h4-5,11,13H,2-3,6H2,1H3
Formula Weight 207.0895
Chemical Name or Material Hydrastinine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.