missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Acetate, TRC

Product Code. 30390518
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30390518 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390518 Supplier Toronto Research Chemicals Supplier No. TRCH68500050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrazine Acetate
CAS 7335-65-1
Molecular Formula C2H8N2O2
Quantity 50 g
Synonym Hydrazine, acetate (1:1), Hydrazine, acetate (7CI), Hydrazine, monoacetate (9CI), Hydrazinium acetate
InChI Formula InChI=1S/C2H4O2.H4N2/c1-2(3)4;1-2/h1H3,(H,3,4);1-2H2
SMILES CC(=O)O.NN
IUPAC Name acetic acid;hydrazine
Molecular Weight (g/mol) 92.1
Formula Weight 92.0586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.