missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.0100 N (N/100), Ricca Chemical

Product Code. 31750254
Change view
Click to view available options
Quantity:
10 L
Packaging:
Cubitainer
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31750254 10 L Cubitainer 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750254 Supplier Ricca Chemical Company Supplier No. R359000010F
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Appearance of Solution Colorless Liquid
CAS Min % 98.0, 0.1
Concentration 0.00995 to 0.01005 N
CAS Max % 100.0, 0.1
Packaging Cubitainer
Quantity 10 L
Synonym hydrochloric acid
Grade Laboratory
Assay Normal at 20°C
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.