missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, ACS, HCl 36.5-38.0%

Product Code. 11397558
Change view
Click to view available options
Quantity:
6 lb.
6 x 6 lb.
Unit Size:
36lb
6lb
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397558 6 lb. 6lb
11307568 6 x 6 lb. 36lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397558 Supplier Thermo Scientific Alfa Aesar Supplier No. 033257.P6

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Acidizing Petroleum Wells, Chemical Intermediate, ore Reduction | CAS: 7647-01-0 | ClH | 36.46 g/mol

Acidizing petroleum wells, chemical intermediate, ore reduction, food processing, pickling and metal cleaning, general cleaning, and laboratory reagent.Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Appearance Colorless pungent
Color Colorless
Density 1.19
Odor Pungent
Assay Percent Range 36.5-38.0%
Quantity 6 lb.
Physical Form Liquid
MDL Number MFCD00011324, MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Formula Weight 36.46
Grade ACS
Chemical Name or Material Hydrochloric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.