missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid solution, 0.1M EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12950854 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12950854 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12950854 Supplier Fisher Chemical Supplier No. VL 017 1000
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
CAS 7647-10-1
Molecular Formula Cl2Pd
Concentration 0.1 M
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Pd++]
IUPAC Name dichloropalladium
Molecular Weight (g/mol) 177.32
PubChem CID 24290
Quantity 1000 mL
ChEBI CHEBI:53434
Grade Eu. Ph. Reference
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.