missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, TRC

Product Code. 30390601
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30390601 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30390601 Supplier Toronto Research Chemicals Supplier No. TRCH9424505G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrofluoric Acid
CAS 7664-39-3
Quantity 5 g
Molecular Formula FH
Synonym Alsurf 45, Anhydrous hydrofluoric Acid, Antisal 2b, Fluorhydric Acid, Fluoric Acid, Fluorine Hydride (FH), Fluorine Monohydride, G-C Hydrofluoride Gel, Hydrofluoric Acid Gas, Hydrogen Fluoride, Hydrogen Fluoride (HF), Hydrogen Monofluoride, Porcelain Conditioner, SA-X, Ultradent Porcelain Etch, Urine Luck 6.3, Vita Ceramic etch
InChI Formula InChI=1S/FH/h1H
SMILES F
IUPAC Name fluorane
Molecular Weight (g/mol) 20.0064
Formula Weight 20.0062
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.