missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Hexachloroplatinate Solution, 8%, Honeywell™ Fluka™

Product Code. 15628290 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 15628290

Brand: Honeywell-Fluka 0066925ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

∼8% in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 16941-12-1
Linear Formula H2PtCl6
MDL Number MFCD00011179
UN Number UN3264
Concentration 8% in H2O
Density 1.060g/mL at 20°C
Formula Weight 409.81g/mol
Percent Purity 8%
Packaging Glass bottle
Quantity 25 mL
Chemical Name or Material Hydrogen Hexachloroplatinate Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.