missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.

Product Code. 11387648
Change view
Click to view available options
Quantity:
50 mL
500 mL
4 x 500 mL
Unit Size:
2000mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387648 50 mL 50mL
11415000 500 mL 500mL
11425000 4 x 500 mL 2000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11387648 Supplier Thermo Scientific Alfa Aesar Supplier No. 033323.AD
  • ONLINE EXCLUSIVE PRICE
    49.37€
    41.65€ / 50mL
    Save 7.72€
    15% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

For Bleaching and Deodorizing Textiles, Wood, Hair | CAS: 7722-84-1 | H2O2 | 34.014 g/mol

For bleaching and deodorizing textiles, wood, hair, etc; rocket propulsion; maturing agent in food; water and sewage treatment; disinfectant; laboratory reagentHydrogen peroxide is used as an oxidizing agent to prepare propylene oxide, dibenzoyl peroxide, lauryl peroxide, peracetic acid and meta-chloroperoxybenzoic acid. It is also involved in the production of inorganic chemicals like sodium perborate and sodium percarbonate, which are used as bleaching agents in detergents. It is an associated iron catalyst and utilized for waste-water treatment to remove organic impurities through an advanced oxidation process. It is also used as a laboratory disinfectant and as a topical antiseptic. In the cosmetic industry, it is used for bleaching human hair and tooth whitening by mixing with ammonium hydroxide and baking soda respectively. Monopropellant engines using hydrogen peroxide as the fuel are commonly used for air-independent underwater propulsion applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Boiling Point 107°C
Density 1.11 g/cm3
Refractive Index 1.335
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Quantity 50 mL
Beilstein 3587191
Physical Form Liquid
Solubility Information Soluble in water,alcohol and ether. Insoluble in petroleumMiscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether.
Formula Weight 34.02
Grade ACS
Chemical Name or Material Hydrogen peroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.