missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab., Thermo Scientific™

For Bleaching and Deodorizing Textiles, Wood, Hair

41.65€ - 55.65€

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387648
SDS View Documents
Thermo Scientific Alfa Aesar
033323.AD
50mL
ONLINE EXCLUSIVE PRICE
45.70€
41.65€ / 50mL
Save 4.05€
8% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11415000
SDS View Documents
Thermo Scientific Alfa Aesar
033323.AP
ONLINE EXCLUSIVE PRICE
61.80€
55.65€ / 500mL
Save 6.15€
9% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7722-84-1
34.014
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide
H2O2
MFCD00011333
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO

Specifications

7722-84-1
MFCD00011333
1.11 g/cm3
Soluble in water,alcohol and ether. Insoluble in petroleumMiscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether.
OO
34.014
CHEBI:16240
ACS
23 hPa (17mm Hg) at 20°C
H2O2
3587191
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
34.02
1.335
Hydrogen peroxide
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P370+P378q-P501c

H271-H302+H332-H314

missing translation for 'dotInformation' : Hazard Class: 5.1, Packing Group: II

missing translation for 'einecsNumber' : 231-765-0

missing translation for 'rtecsNumber' : MX0899000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.