Learn More
VEE GEE Hydrometer Jar
Product Code. 17806483 Shop All Thermo Scientific Incubateurs à CO2 Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
600 mL
1000 mL
Includes:
Graduation Line
No Graduation Line
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes unitSize
17806483 1000 mL Graduation Line Each
17806523 100 mL No Graduation Line Each
17866573 250 mL No Graduation Line Each
17846573 1000 mL No Graduation Line Each
17816583 500 mL No Graduation Line Each
17816483 600 mL No Graduation Line Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17806483 Supplier VEE GEE Supplier No. 210501000L
Only null left
Add to basket
Add to basket
The Hydrometer Jar is made of borosilicate glass, the jar features a beaded rim and a circular base. Sized properly to eliminate surface tension effects that lead to inaccurate readings.
  • The capacity varies depending on the model.
Especificaciones
Diameter (Metric) 15 cm
Capacity (Metric) 1000 mL
Material Borosilicate glass
Disposable Reusable
Includes Graduation Line
Base Type Flat Circular
Capacity (English) 34 fl. oz.
Height (English) 16 in.
Height (Metric) 40 cm
Product Type Hydrometer Jar
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.