missing translation for 'onlineSavingsMsg'
Learn More

Hydroxocobalamin (>90%), TRC

Product Code. 30005593
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005593 250 mg 250mg
30007062 1 g 1g
17969914 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005593 Supplier Toronto Research Chemicals Supplier No. TRCH862820250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13422-51-0
Molecular Formula C62 H89 Co N13 O15 P
Molecular Weight (g/mol) 1346.36
Synonym Cobinamide, Co-hydroxy-, f-(dihydrogen phosphate), inner salt, 3'-ester with (5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole-κN3), Cobinamide, dihydroxide, dihydrogen phosphate (ester), mono(inner salt), 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole, Cobinamide, dihydroxide, dihydrogen phosphate (ester), mono(inner salt), 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole, Alpha Cobione, AlphaRedisol, Axlon, Ciplamin H, Cobalamin, hydroxo-, Cobalex, Cobalin H, Cobinamide hydroxide phosphate 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole inner salt, Codroxomin, Docclan, Docelan, Docevita, Droxomin, Ducobee Hy, Duradoce, Duralta-12, Hydrocobalamin, Hydrogrisevit, Hydrovit, Hydroxocobalamin, Hydroxocobalamine, Hydroxocobemine, Hydroxy vitamin B12, Hydroxycobalamin, Hydroxycobalamine, Hyxobamine, Idrogrisevit, Megamilbedoce, Neo-Betalin 12, Neo-Cytamen, Neo-Macrabin, Neo-Rojamin, OH-Duphar, Oxobemin, Primabalt RP, Redisol H, Sytobex H, Vibeden, Vitadurin, Vitamin B12a, α-(5,6-Dimethylbenzimidazolyl)hydroxocobamide
SMILES C[C@@H]1CNC(=O)CC[C@]2(C)[C@@H](CC(=O)N)[C@H]3N4C2=C(C)C5=N6C(=CC7=N8C(=C(C)C9=N([C@]3(C)[C@@](C)(CC(=O)N)[C@@H]9CCC(=O)N)[Co+]468(O)N%10=CN([C@H]%11O[C@H](CO)[C@@H](OP(=O)([O-])O1)[C@H]%11O)c%12cc(C)c(C)cc%10%12)[C@@](C)(CC(=O)N)[C@@H]7CCC(=O)N)C(C)(C)[C@@H]5CCC(=O)N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C62H90N13O14P.Co.H2O/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;1H2/q;+3;/p-3/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57+,59-,60+,61+,62+;;/m1../s1
Formula Weight 1345.5671
Chemical Name or Material Hydroxocobalamin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.