missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Bupropion, TRC

Product Code. 17992995
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992995 5 mg 5mg
17913955 10 mg 10mg
17911595 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992995 Supplier Toronto Research Chemicals Supplier No. TRCH8306755MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357399-43-0
Molecular Formula C13 H18 Cl N O2
Molecular Weight (g/mol) 255.74
Synonym 2-Morpholinol, 2-(3-chlorophenyl)-3,5,5-trimethyl-, 2-(3-Chlorophenyl)-3,5,5-trimethyl-2-morpholinol, 2-Hydroxy-2-(3-chlorophenyl)-3,5,5-trimethylmorpholine, Hydroxy Bupropion, Bupropion Morpholinol, Bupropion-morpholinol
IUPAC Name 2-(3-chlorophenyl)-3,5,5-trimethylmorpholin-2-ol
SMILES CC1NC(C)(C)COC1(O)c2cccc(Cl)c2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H18ClNO2/c1-9-13(16,17-8-12(2,3)15-9)10-5-4-6-11(14)7-10/h4-7,9,15-16H,8H2,1-3H3
Formula Weight 255.1026
Percent Purity >95%
Chemical Name or Material Hydroxy Bupropion
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.