missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Celecoxib, TRC

Product Code. 30397619
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30397619 100 mg 100mg
30396952 10 mg -
30391570 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397619 Supplier Toronto Research Chemicals Supplier No. TRCH946580100MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170571-00-3
Molecular Formula C17 H14 F3 N3 O3 S
Molecular Weight (g/mol) 397.37
Synonym Benzenesulfonamide, 4-[5-[4-(hydroxymethyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-1-yl]- (9CI, ACI), 4-[5-[4-(Hydroxymethyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide (ACI), 4-[5-[4-(Hydroxymethyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide, Hydroxymethylcelecoxib, Hydroxymethyl Celecoxib, Hydroxy Celecoxib
IUPAC Name 4-[5-[4-(hydroxymethyl)phenyl]-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
SMILES NS(=O)(=O)c1ccc(cc1)n2nc(cc2c3ccc(CO)cc3)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H14F3N3O3S/c18-17(19,20)16-9-15(12-3-1-11(10-24)2-4-12)23(22-16)13-5-7-14(8-6-13)27(21,25)26/h1-9,24H,10H2,(H2,21,25,26)
Formula Weight 397.0708
Percent Purity >95%
Chemical Name or Material Hydroxymethylcelecoxib (4-[5-[4-(Hydroxymethyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.