missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine N-Oxide, TRC

Product Code. 30136212
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136212 50 mg 50mg
30011682 10 mg 10mg
30005822 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136212 Supplier Toronto Research Chemicals Supplier No. TRCH90531050MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449223-88-4
Molecular Formula C18 H26 Cl N3 O2
Molecular Weight (g/mol) 351.87
Synonym Hydroxychloroquine Impurity B
IUPAC Name 4-[(7-chloroquinolin-4-yl)amino]-N-ethyl-N-(2-hydroxyethyl)pentan-1-amine oxide
SMILES CC[N+]([O-])(CCO)CCCC(C)Nc1ccnc2cc(Cl)ccc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H26ClN3O2/c1-3-22(24,11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21)
Formula Weight 351.1714
Percent Purity >95%
Chemical Name or Material Hydroxychloroquine N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.