missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine, TRC

Product Code. 30011666
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011666 50 mg 50mg
30005722 100 mg 100mg
30005261 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011666 Supplier Toronto Research Chemicals Supplier No. TRCB41975050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-42-3
Molecular Formula C18 H26 Cl N3 O
Molecular Weight (g/mol) 335.87
Synonym Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]ethylamino]-, Ethanol, 2-[[4-[(7-chloro-4-quinolyl)amino]pentyl]ethylamino]- (6CI,8CI), 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]ethylamino]ethanol, (±)-Hydroxychloroquine, 7-Chloro-4-[4-(N-ethyl-N-β-hydroxyethylamino)-1-methylbutylamino]quinoline, 7-Chloro-4-[4-[ethyl(2-hydroxyethyl)amino]-1-methylbutylamino]quinoline, 7-Chloro-4-[5-(N-ethyl-N-2-hydroxyethylamino)-2-pentyl]aminoquinoline, Hydroxychloroquine, Oxichloroquine, Oxychlorochin, Oxychloroquine, Racemic Hydroxychloroquine, WIN 1258, Win 1258-2
IUPAC Name 2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol
SMILES CCN(CCO)CCCC(C)Nc1ccnc2cc(Cl)ccc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H26ClN3O/c1-3-22(11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21)
Formula Weight 335.1764
Percent Purity >95%
Chemical Name or Material Hydroxychloroquine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.