missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine sulfate, 99%

Product Code. 10286920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
1kg
5g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10286920 5 g Glass Bottle 5g
10701861 100 g Plastic bottle 100g
10722051 1 kg Plastic bottle 1kg
10464025 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10286920 Supplier Thermo Scientific Acros Supplier No. 198530050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 10039-54-0 | H6N2O2·H2SO4 | 164.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10039-54-0
Molecular Formula H6N2O2·H2SO4
MDL Number MFCD00044869
Synonym Hydroxylammonium sulfate

Specifications

Color White
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99%
Linear Formula (H2NOH)2·H2SO4
Fieser 01,481
Merck Index 15, 4867
Formula Weight 164.14
Percent Purity ≥98.5%
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Hydroxylamine sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.