Learn More
Ansell™ HyFlex 11-944 Gloves
Description
- An ideal alternative to light leather gloves.
- Fine and stretchable stitches for excellent comfort.
- Designed specifically for use in moderately oily manufacturing and maintenance environments where protection combined with abrasion resistance are essential.
Specifications
Specifications
| Coating Material | Nitrile |
| Color | White, Black |
| Cut Resistance Rating | ANSI A1 |
| Exterior Finish | No Texture |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Nylon |
| Product Type | Coated Gloves |
| Size | 7 |
| Glove Cuff Style | Knit |
| Show More |