missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Hygromycin B ReadyMade™ Solution

Product Code. 30044054
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
5 x 20 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044054 5 x 20 mL 5mL
30044015 20 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044054 Supplier TOKU-E™ Supplier No. H0115x20ML
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hygromycin B ReadyMade™ Solution is provided as a sterile-filtered solution in PBS buffer.

  • CAS Number: 31282-04-9
  • Molecular Formula: C20H37N3O13
  • Molecular Weight: 527.52 g/mol
  • Storage Conditions: 2-8°C
  • Form: Liquid
  • Source: Streptomyces hygroscopicus
  • Spectrum: Hygromycin B is effective against bacteria, fungi, and higher eukaryotic cells
  • Mechanism of Action: Hygromycin B inhibits protein synthesis by strengthening the interaction of tRNA binding in the ribosomal A-site. Hygromycin B also prevents mRNA and tRNA translocation by an unknown mechanism. These are unique mechanisms for an aminoglycoside antibiotic and they differ from the mode of action of neomycin, gentamicin, and G418

Specifications

CAS 31282-04-9
Certifications/Compliance Purity >90% (HPLC)
Chemical Name or Material Hygromycin B
Color Clear, yellowish
Content And Storage 2°C to 8°C
Description Hygromycin B ReadyMade™ Solution 50 mg/mL in PBS buffer
Concentration 50 mg/mL
Form Clear, colorless to slight yellow solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in Phosphate-buffered saline (PBS)
For Use With (Application) Microbiology, Plant Biololgy, Eukaryotic Cell Culture Application
Product Line ReadyMade™ Solutions
Purity or Quality Grade >90%
Quantity 5 x 20 mL
Shipping Condition Ambient
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.